Coenzyme Q10 ≥98%
Supplier: Thermo Scientific Chemicals
J65137-03
J65137-06
J65137-14
CAAAJ65137-03EA
129.4
CAD
CAAAJ65137-03
CAAAJ65137-06
CAAAJ65137-14
Coenzyme Q10 ≥98%
Coenzyme Q10
Component of electron transport chain and in aerobic cellular respiration
Powder
Applications: Component of electron transport chain and in aerobic cellular respiration
Formula:
C₅₉H₉₀O₄ Melting Pt: 48…52 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042919 CAS Number: 303-98-0 EINECS: 206-147-9 Merck Index: 14,9843 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...