3'-Azido-3'-deoxythymidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine, Azidothymidine, AZT

J65127-03 J65127-06 J65127-14
CAAAJ65127-03EA 82 CAD
CAAAJ65127-03 CAAAJ65127-06 CAAAJ65127-14
3'-Azido-3'-deoxythymidine 98%
3'-Azido-3'-deoxythymidine

Anti-viral agent


Powder


Applications: Anti-viral agent

Formula: C₁₀H₁₃N₅O₄
MW: 267.25 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006536
CAS Number: 30516-87-1
Merck Index: 14,10123

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR