ɑ-Asarone

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Asarone, trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene, (E)-1,2,4-Trimethoxy-5-(prop-1-en-1-yl)benzene

J65125-03 J65125-06
CAAAJ65125-03EA 101.12 CAD
CAAAJ65125-03 CAAAJ65125-06
ɑ-Asarone
alpha-Asarone

Powder


CAS No.: 2883-98-9 Molecular Formula: C12H16O3 Formula Weight: 208.25 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to yellow Melting Point: 57-61° Boiling Point: 296° Beilstein Reference: 1910606 MDL No.: MFCD00064457

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 296 °C
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00064457
CAS Number: 2883-98-9
EINECS: 220-743-6
Merck Index: 14,824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR