1-Aminocyclopropanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

J65118-03 J65118-MD
CAAAJ65118-03EA 273.34 CAD
CAAAJ65118-03 CAAAJ65118-MD
1-Aminocyclopropanecarboxylic acid 98%
1-Aminocyclopropanecarboxylic acid

A NMDA glutamate receptor agonist at glycine site


Crystalline powder

Formula: C₄H₇NO₂
Melting Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00009944
CAS Number: 22059-21-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR