Guanidinium thiocyanate for molecular biology

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

J65104-18 J65104-30 J65104-36
CAAAJ65104-18EA 153.58 CAD
CAAAJ65104-18 CAAAJ65104-30 CAAAJ65104-36
Guanidinium thiocyanate for molecular biology
Guanidinium thiocyanate

A protein denaturant


Crystalline solid

Formula: NH₂C(=NH)NH₂·HSCN
Melting Pt: 118…122 °C
Density: 1.29
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR