Nɑ-Acetyl-L-lysine 99%
Supplier: Thermo Scientific Chemicals
J65083-03
J65083-06
CAAAJ65083-03EA
120.87
CAD
CAAAJ65083-03
CAAAJ65083-06
Nɑ-Acetyl-L-lysine 99%
Nalpha-Acetyl-L-lysine
Powder
Purity: 99%
CAS No.: 1946-82-3
Molecular Formula: C8H16N2O3
Formula Weight: 188.20
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 256-258° dec.
MDL No.: MFCD00008233
Formula:
C₈H₁₆N₂O₃ Melting Pt: 256…258 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008233 CAS Number: 1946-82-3 EINECS: 217-747-5 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...