Biotin-PEG3-azide
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]biotinamide, Azido-PEG3-biotin conjugate
J64996-MB
J64996-MC
CAAAJ64996-MBEA
267.02
CAD
CAAAJ64996-MB
CAAAJ64996-MC
Biotin-PEG3-azide
Biotin-PEG3-azide
An azide-biotin conjugate with PEG spacer to improve solubility in aqueous media
Solid
Applications: An azide-biotin conjugate with PEG spacer to improve solubility in aqueous media
Formula:
C₁₈H₃₂N₆O₅S MW: 444.55 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20134145 CAS Number: 875770-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...