O-Acetyl-L-serine hydrochloride

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-3-Acetoxy-2-aminopropanoic acid hydrochloride, H-Ser(Ac)-OH.HCl

J64933-03 J64933-06
CAAAJ64933-03EA 127.66 CAD
CAAAJ64933-03 CAAAJ64933-06
O-Acetyl-L-serine hydrochloride
O-Acetyl-L-serine hydrochloride

Substrate for cysteine synthase and beta-substituted alanine synthase in higher plants


Powder


Applications: Substrate for cysteine synthase and beta-substituted alanine synthase in higher plants

Formula: C₅H₉NO₄
MW: 183.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060169
CAS Number: 66638-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR