2-Aminopurine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Purin-2-amine, 7H-Purin-2-amine

J64919-03 J64919-MD J64919-ME
CAAAJ64919-03EA 840.56 CAD
CAAAJ64919-03 CAAAJ64919-MD CAAAJ64919-ME
2-Aminopurine 98%
2-Aminopurine

A protein kinase R inhibitor


Powder

Formula: C₅H₅N₅
Melting Pt: 280…282 °C
Storage Temperature: Ambient
MDL Number: MFCD00005566
CAS Number: 452-06-2
EINECS: 207-197-4

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR