N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64902-MB
J64902-MC
CAAAJ64902-MBEA
200.66
CAD
CAAAJ64902-MB
CAAAJ64902-MC
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
Labeling reagents with enhanced water solubility and amine reactivity as the activated NHS ester
Molecular Formula: C18H27NO9
Formula Weight: 401.41
Storage Temperature: -30°C to -10°C
Physical Form: Viscous liquid
Appearance: Pale yellow to orange
MDL No.: MFCD22380743
Applications: Labeling reagents with enhanced water solubility and amine reactivity as the activated NHS ester
Formula:
C₁₈H₂₇NO₉ MW: 401.41 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22380743 CAS Number: 1428629-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...