Benzopurpurine 4B

Supplier: Thermo Scientific Chemicals

J64901-14 J64901-22
CAAAJ64901-14EA 97.01 CAD
CAAAJ64901-14 CAAAJ64901-22
Benzopurpurine 4B
Benzopurpurine 4B

Powder


CAS No.: 992-59-6 Molecular Formula: C34H26N6Na2O6S2 Formula Weight: 724.73 Physical Form: Powder Appearance: Dark red MDL No.: MFCD00012410

Formula: C₃₄H₂₈N₆O₆S₂
Storage Temperature: Ambient
MDL Number: MFCD00012410
CAS Number: 992-59-6
EINECS: 213-594-3
Merck Index: 14,1011

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR