Acetylene-PEG4-maleimide
Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(N-maleimidyl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propionamide
J64859-MB J64859-MC
CAAAJ64859-MBEA 232.26 CAD
CAAAJ64859-MB CAAAJ64859-MC CAAAJ64859-ME
Acetylene-PEG4-maleimide
Acetylene-PEG4-maleimide
A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility
Order Now
SPECIFICATIONS
Environmentally Preferable