Diphenyleneiodonium chloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64838-MC
J64838-MF
CAAAJ64838-MCEA
831.08
CAD
CAAAJ64838-MC
CAAAJ64838-MF
Diphenyleneiodonium chloride
Diphenyleneiodonium chloride
Powder
CAS: 4673-26-1
MDL: MFCD00214165
Formula:
C₁₂H₈ClI Melting Pt: >310 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00214165 CAS Number: 4673-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...