Azido-PEG4-NHS ester
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Succinimidyl 15-Azido-4,7,10,13-tetraoxapentadecanoate, NHS-PEG4-Azide, 15-Azido-4,7,10,13-tetraoxapentadecanoic acid N-succinimidyl ester
An amine reactive reagent for azide incorporation. Contains a PEG spacer
Molecular Formula: C15H24N4O8
Formula Weight: 308.37
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Colorless to pale yellow
MDL No.: MFCD13184948
Applications: An amine reactive reagent for azide incorporation. Contains a PEG spacer
Formula:
C₁₅H₂₄N₄O₈ MW: 388.38 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13184948 CAS Number: 944251-24-5 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...