Bismarck brown R

Supplier: Thermo Scientific Chemicals

J64757-14 J64757-22
CAAAJ64757-14EA 55.62 CAD
CAAAJ64757-14 CAAAJ64757-22
Bismarck brown R
Bismarck Brown R

Powder


CAS: 5421-66-9 MDL: MFCD00036386

Formula: C₂₁H₂₄N₈·2HCl
Melting Pt: 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00036386
CAS Number: 5421-66-9
EINECS: 226-541-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR