Choline hydrogen tartrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

J64657-30 J64657-A1 J64657-A4
CAAAJ64657-30EA 75.52 CAD
CAAAJ64657-30 CAAAJ64657-A1 CAAAJ64657-A4
Choline hydrogen tartrate 98+%
Choline hydrogen tartrate

Powder


CAS: 87-67-2 MDL: MFCD00036332

Formula: C₉H₁₉NO₇
MW: 253.25 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00036332
CAS Number: 87-67-2
EINECS: 201-763-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR