Benserazide hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benserazide HCI, DL-Serine 2-(2,3,4-Trihydroxybenzyl)hydrazide Hydrochloride, 2-Amino-3-hydroxy-N'-(2,3,4-trihydroxybenzyl)propanehydrazide hydrochloride

J64581-03 J64581-06
CAAAJ64581-03EA 148.52 CAD
CAAAJ64581-03 CAAAJ64581-06
Benserazide hydrochloride
Benserazide hydrochloride

Inhibitor of L-aromatic amino acid decarboxylase


Powder


Applications: Inhibitor of L-aromatic amino acid decarboxylase

Formula: C₁₀H₁₅N₃O₅
MW: 293.70 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00078571
CAS Number: 14919-77-8
EINECS: 238-991-9
Merck Index: 14,1050

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR