Cinnarizine

Supplier: Thermo Scientific Chemicals

J64568-14 J64568-22
CAAAJ64568-14EA 142.83 CAD
CAAAJ64568-14 CAAAJ64568-22
Cinnarizine
Cinnarizine

An antihistamine


Powder


Applications: An antihistamine

Formula: C₂₆H₂₈N₂
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00056037
CAS Number: 298-57-7
EINECS: 206-064-8
Merck Index: 14,2305

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR