1-Amino-3,6,9,12-tetraoxapentadec-14-yne

Supplier: Thermo Scientific Chemicals
Warning

J64527-MC J64527-ME
CAAAJ64527-MCEA 605.14 CAD
CAAAJ64527-MC CAAAJ64527-ME
1-Amino-3,6,9,12-tetraoxapentadec-14-yne
1-Amino-3,6,9,12-tetraoxapentadec-14-yne

Acetylene containing reagent with PEG spacer and carboxyl reactivity

Formula: C₁₁H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD22380736
CAS Number: 1013921-36-2
EINECS: 852-311-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR