PD 98059 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one

J64485-MA J64485-MB
CAAAJ64485-MAEA 211.72 CAD
CAAAJ64485-MA CAAAJ64485-MB
PD 98059 99%
PD 98059

A MAP kinase inhibitor


Purity: 99% CAS No.: 167869-21-8 Molecular Formula: C16H13NO3 Formula Weight: 267.28 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Yellow MDL No.: MFCD00671789


Applications: A MAP kinase inhibitor

Formula: C₁₆H₁₃NO₃
MW: 267.284 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00671789
CAS Number: 167869-21-8

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR