5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

J64451-03 J64451-MD
CAAAJ64451-03EA 448.72 CAD
CAAAJ64451-03 CAAAJ64451-MD
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Substrate for alkaline phosphatase


Powder

Formula: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR