Sodium-4-aminosalicylate dihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

J64449-14 J64449-22 J64449-36
CAAAJ64449-14EA 42.82 CAD
CAAAJ64449-14 CAAAJ64449-22 CAAAJ64449-36
Sodium-4-aminosalicylate dihydrate 98%
Sodium 4-aminosalicylate dihydrate

Powder


CAS: 6018-19-5 MDL: MFCD00151044

Formula: C₇H₆NNaO₃·2H₂O
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00064392
CAS Number: 6018-19-5
EINECS: 205-091-2

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR