Carbetapentane citrate, white ≥95% (by HPLC), powder

Supplier: Thermo Scientific Chemicals

J64446-03 J64446-MF
CAAAJ64446-03EA 333.38 CAD
CAAAJ64446-03 CAAAJ64446-MF
Carbetapentane citrate, white ≥95% (by HPLC), powder
Carbetapentane citrate

Sigma receptor agonist


Powder


Applications: Sigma receptor agonist

Formula: C₂₆H₃₉NO₁₀
Storage Temperature: Ambient
MDL Number: MFCD00055697
CAS Number: 23142-01-0
EINECS: 245-449-5
Merck Index: 14,1794

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR