5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt 98%

Supplier: Thermo Scientific Chemicals

Synonyms: XGLUC Sodium Salt, 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt

J64360-03 J64360-MD
CAAAJ64360-03EA 581.44 CAD
CAAAJ64360-03 CAAAJ64360-MD
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt 98%
5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide sodium salt

Substrate for beta-glucuronidase


Powder


Applications: Substrate for beta-glucuronidase

Formula: C₁₄H₁₃BrClNO₇
MW: 444.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00135782
CAS Number: 129541-41-9

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR