4-Amino-1,8-naphthalimide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64358-03
J64358-MC
CAAAJ64358-03EA
346.02
CAD
CAAAJ64358-03
CAAAJ64358-MC
4-Amino-1,8-naphthalimide 95%
4-Amino-1,8-naphthalimide
Solid
Purity: 95%
CAS: 1742-95-6
MDL: MFCD00006921
Formula:
C₁₂H₈N₂O₂ Boiling Pt: 518 °C (1013 hPa) Melting Pt: >360 °C Density: 1.105 Storage Temperature: Refrigerator |
MDL Number:
MFCD00006921 CAS Number: 1742-95-6 EINECS: 217-110-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...