Amodiaquine dihydrochloride dihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Amodiaquin dihydrochloride dihydrate, 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 4-([7-Chloro-4-quinolinyl]amino)-2-([diethylamino]methyl)phenol, 4-[(7-Chloroquinolin-4-yl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride

J64326-06 J64326-14
CAAAJ64326-06EA 54.04 CAD
CAAAJ64326-06 CAAAJ64326-14 CAAAJ64326-22
Amodiaquine dihydrochloride dihydrate 98%
Amodiaquine dihydrochloride dihydrate

An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor


Yellow powder


Applications: An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor

Formula: C20H22ClN3O .2HCl .2H2O
MW: 464.812 g/mol (392.33 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00078857
CAS Number: 6398-98-7
EINECS: 200-706-0
Merck Index: 14,572

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR