Cloxacillin sodium salt

Supplier: Thermo Scientific Chemicals

J64314-03 J64314-06 J64314-14
CAAAJ64314-03EA 89.27 CAD
CAAAJ64314-03 CAAAJ64314-06 CAAAJ64314-14
Cloxacillin sodium salt
Cloxacillin sodium salt

A semisynthetic antibiotic


Powder


Applications: A semisynthetic antibiotic

Formula: C₁₉H₁₇ClN₃NaO₅S
Melting Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00063568
CAS Number: 642-78-4
EINECS: 211-390-9
Merck Index: 14,2419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR