Meclofenoxate hydrochloride 98%, powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Dimethylamino)ethyl 2-(4-Chlorophenoxy)acetate Hydrochloride, 2-(4-Chlorophenoxy)acetic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride, Centrophenoxine hydrochloride

J64196-06 J64196-14 J64196-22
CAAAJ64196-06EA 73.63 CAD
CAAAJ64196-06 CAAAJ64196-14 CAAAJ64196-22
Meclofenoxate hydrochloride 98%, powder
Meclofenoxate hydrochloride

Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats


Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid.


Applications: Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats

Formula: C₁₂H₁₆ClNO₃
MW: 294.17 g/mol
Melting Pt: 135…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00012533
CAS Number: 3685-84-5
EINECS: 222-975-3
Merck Index: 14,5780

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR