5-Azacytidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ladakamycin, 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one
J64159-03
J64159-06
CAAAJ64159-03EA
117.55
CAD
CAAAJ64159-03
CAAAJ64159-06
5-Azacytidine 98%
5-Azacytidine
Causes DNA demethylation or hemi-demethylation; a powerful mutagen
Powder
Applications: Causes DNA demethylation or hemi-demethylation; a powerful mutagen
Formula:
C₈H₁₂N₄O₅ MW: 244.21 g/mol Melting Pt: 226…232 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006539 CAS Number: 320-67-2 EINECS: 206-280-2 Merck Index: 14,890 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...