Acyclovir 98%

Supplier: Thermo Scientific Chemicals
Warning

J64144-03 J64144-06
CAAAJ64144-03EA 132.56 CAD
CAAAJ64144-03 CAAAJ64144-06 CAAAJ64144-14
Acyclovir 98%
Acyclovir

Inhibits cytomegalovirus replication; induces apoptosis


Powder


Applications: Inhibits cytomegalovirus replication; induces apoptosis

Formula: C₈H₁₁N₅O₃
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00057880
CAS Number: 59277-89-3
EINECS: 261-685-1
Merck Index: 14,146

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR