Chitosan ≥85%, deacetylated

Supplier: Thermo Scientific Chemicals

Synonyms: Deacetylated chitin, Poly(D-glucosamine)

J64143-18 J64143-30 J64143-36
CAAAJ64143-18EA 97.64 CAD
CAAAJ64143-18 CAAAJ64143-30 CAAAJ64143-36
Chitosan ≥85%, deacetylated
Chitosan

Chitosan is used as a flocculant. It also finds use in protein precipitation and as an encapsulating agent.


Powder


Applications: Chitosan is used as a flocculant. It also finds use in protein precipitation and as an encapsulating agent.

Storage Temperature: Ambient
MDL Number: MFCD00161512
CAS Number: 9012-76-4

Order Now

Specification Test Results

Assay Minimum 85


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR