Nε-acetyl-L-lysine 99%

Supplier: Thermo Scientific Chemicals

J64139-03 J64139-06
CAAAJ64139-03EA 105.39 CAD
CAAAJ64139-03 CAAAJ64139-06
Nε-acetyl-L-lysine 99%
Nepsilon-acetyl-L-lysine

Powder


Purity: 99% CAS No.: 692-04-6 Molecular Formula: C8H16N2O3 Formula Weight: 188.20 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 250° dec. MDL No.: MFCD00002639

Formula: C₈H₁₆N₂O₃
Melting Pt: 250 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002639
CAS Number: 692-04-6
EINECS: 211-725-9

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR