N(α)-Acetyl-L-tryptophan 99%
Supplier: Thermo Scientific Chemicals
J64096-06
J64096-14
CAAAJ64096-06EA
57.67
CAD
CAAAJ64096-06
CAAAJ64096-14
CAAAJ64096-22
N(α)-Acetyl-L-tryptophan 99%
N(α)-Acetyl-L-tryptophan
Powder
CAS: 1218-34-4
MDL: MFCD00065976
Formula:
C₁₃H₁₄N₂O₃ Melting Pt: 186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065976 CAS Number: 1218-34-4 EINECS: 214-935-9 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...