(S)-(+)-2-Amino-4-phosphonobutyric acid 97%

Supplier: Thermo Scientific Chemicals
Warning

J64075-MA J64075-MCR
CAAAJ64075-MAEA 413.96 CAD
CAAAJ64075-MA CAAAJ64075-MCR
(S)-(+)-2-Amino-4-phosphonobutyric acid 97%
(S)-(+)-2-Amino-4-phosphonobutyric acid

A NMDA agonist


Powder


Applications: A NMDA agonist

Formula: C₄H₁₀NO₅P
Storage Temperature: Ambient
MDL Number: MFCD00083244
CAS Number: 23052-81-5

Order Now

Specification Test Results

Assay Minimum 99

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR