Imiquimod 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine, 4-Amino-1-isobutyl-1H-imidazo[4,5-c]quinoline

J63990-03 J63990-MC
CAAAJ63990-03EA 628.84 CAD
CAAAJ63990-03 CAAAJ63990-MC
Imiquimod 99%
Imiquimod

A caspase 3 activator that acts as an immunomodulator and displays antiviral and antitumor activity


Solid


Applications: A caspase 3 activator that acts as an immunomodulator and displays antiviral and antitumor activity

Formula: C₁₄H₁₆N₄
MW: 240.31 g/mol
Melting Pt: 292…294 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866946
CAS Number: 99011-02-6
UN: 2811
ADR: 6.1 III
Merck Index: 14,4922

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR