Lofexidine hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J63960-03
J63960-06
CAAAJ63960-03EA
252.8
CAD
CAAAJ63960-03
CAAAJ63960-06
Lofexidine hydrochloride ≥98%
Lofexidine hydrochloride
Alpha-2 adrenergic agonist
Crystalline powder
Applications: Alpha-2 adrenergic agonist
Formula:
C₁₁H₁₃Cl₃N₂O Melting Pt: 225…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00917022 CAS Number: 21498-08-8 UN: 2811 ADR: "6.1",III Merck Index: 14,5558 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...