Coomassie brilliant blue G-250 solution ready-to-use
Supplier: Thermo Scientific Chemicals
J63797-AP
J63797-K2
CAAAJ63797-APEA
142.83
CAD
CAAAJ63797-AP
CAAAJ63797-K2
Coomassie brilliant blue G-250 solution ready-to-use
Coomassie brilliant blue G-250
This solution contains 30% methanol, 5% acetic acid, 0.2% Brilliant Blue G.
CAS No.: 6104-58-1
Molecular Formula: C47H48N3NaO7S2
Formula Weight: 854.04
Physical Form: Liquid
Appearance: Dark blue
Beilstein Reference: 5230822
UN No.: 1992
MDL No.: MFCD00078482
Formula:
C₄₇H₄₈N₃NaO₇S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00078482 CAS Number: 6104-58-1 EINECS: 228-058-4 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...