Sodium hydroxide 10 mol/l (10 N) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

J63736-AE J63736-AK
CAAAJ63736-AEEA 96.57 CAD
CAAAJ63736-AE CAAAJ63736-AK
Sodium hydroxide 10 mol/l (10 N) in aqueous solution
Sodium hydroxide

Liquid


CAS: 1310-73-2 MDL: MFCD00003548

Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: Max. 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR