N,N,N',N'-Tetramethylethylenediamine (TEMED) for electrophoresis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine

J63734-AC J63734-AE
CAAAJ63734-ACEA 90.49 CAD
CAAAJ63734-AC CAAAJ63734-AE
N,N,N',N'-Tetramethylethylenediamine (TEMED) for electrophoresis
N,N,N',N'-Tetramethylethylenediamine (TEMED)

Liquid


CAS: 110-18-9 MDL: MFCD00008335

Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 120
Melting Pt: -64
Density: 0.775
Flash Pt: 18 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3 II
Merck Index: 14,9134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR