Nimesulide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide, N-(4-Nitro-2-phenoxyphenyl)methanesulphonamide

J63699-06 J63699-14
CAAAJ63699-06EA 94.33 CAD
CAAAJ63699-06 CAAAJ63699-14
Nimesulide
Nimesulide

Highly selective cyclooxygenase-2 inhibitor which induces apoptosis


Needle-like crystals


Applications: Highly selective cyclooxygenase-2 inhibitor which induces apoptosis

Formula: C₁₃H₁₂N₂O₅S
MW: 308.308 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079470
CAS Number: 51803-78-2
EINECS: 257-431-4
UN: 2811
ADR: 6.1 III
Merck Index: 14,6548

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR