Elantan 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J63674-06
J63674-09
J63674-14
CAAAJ63674-06EA
333.38
CAD
CAAAJ63674-06
CAAAJ63674-09
CAAAJ63674-14
Elantan 98+%
Elantan
A coronary vasodilator that does not require hepatic biotransformation
Crystalline powder
Applications: A coronary vasodilator that does not require hepatic biotransformation
Formula:
C₆H₉NO₆ Melting Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143462 CAS Number: 16051-77-7 EINECS: 240-197-2 UN: 3251 ADR: "4.1",III Merck Index: 14,5225 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...