Chlorpromazine hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

J63659-09 J63659-14 J63659-22
CAAAJ63659-09EA 156.89 CAD
CAAAJ63659-09 CAAAJ63659-14 CAAAJ63659-22
Chlorpromazine hydrochloride ≥98%
Chlorpromazine hydrochloride

Reported to be useful as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.


Crystalline solid


Applications: Reported to be useful as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.

Formula: C₁₇H₁₉ClN₂S
MW: 355.32 g/mol
Melting Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00012654
CAS Number: 69-09-0
EINECS: 200-701-3
UN: 2811
ADR: "6.1",III
Merck Index: 14,2185

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR