Dicoumarol

Supplier: Thermo Scientific Chemicals
Danger

J63634-06 J63634-14
CAAAJ63634-06EA 63.83 CAD
CAAAJ63634-06 CAAAJ63634-14
Dicoumarol
Dicoumarol

Vitamin K reductase inhibitor possessing anticoagulant properties


Powder


Applications: Vitamin K reductase inhibitor possessing anticoagulant properties

Formula: C₁₉H₁₂O₆
Melting Pt: 286…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00006857
CAS Number: 66-76-2
EINECS: 200-632-9
UN: 2811
ADR: 6.1,III
Merck Index: 14,3090

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR