Acemetacin

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: [1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetoxyacetic Acid, 2-(2-(1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetoxy)acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic Acid Carboxymethyl Ester

J63583-04 J63583-09
CAAAJ63583-04EA 108.7 CAD
CAAAJ63583-04 CAAAJ63583-09
Acemetacin
Acemetacin

A potent inhibitor of COX-2 found to have anti-tumor activity in the colon


Powder


Applications: A potent inhibitor of COX-2 found to have anti-tumor activity in the colon

Formula: C₂₁H₁₈ClNO₆
MW: 415.83 g/mol
Melting Pt: 149…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151473
CAS Number: 53164-05-9
EINECS: 258-403-4
UN: 2811
ADR: 6.1 III
Merck Index: 14,27

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR