Honokiol ≥98%

Supplier: Thermo Scientific Chemicals
Danger

J63434-MA J63434-MB J63434-MC
CAAAJ63434-MAEA 205.4 CAD
CAAAJ63434-MA CAAAJ63434-MB CAAAJ63434-MC
Honokiol ≥98%
Honokiol

Induces apoptosis in human squamous lung cancer cells


Powder


Applications: Induces apoptosis in human squamous lung cancer cells

Formula: C₁₈H₁₈O₂
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00016674
CAS Number: 35354-74-6
UN: 3077
ADR: "9",III
Merck Index: 14,4742

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR