Tetrahydropalmatine 98%

Supplier: Thermo Scientific Chemicals

J63328-03 J63328-06
CAAAJ63328-03EA 110.44 CAD
CAAAJ63328-03 CAAAJ63328-06
Tetrahydropalmatine 98%
Tetrahydropalmatine

Powder


Purity: 98% CAS No.: 2934-97-6 Molecular Formula: C21H25NO4 Formula Weight: 355.43 Physical Form: Powder Appearance: Pale yellow Melting Point: 155° MDL No.: MFCD00214191

Formula: C₂₁H₂₅NO₄
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00214191
CAS Number: 2934-97-6

Order Now

Specification Test Results

Assay Minimum 98

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR