Bisacrylamide 2% in aqueous solution

Supplier: Thermo Scientific Chemicals

J63265-AP J63265-K2
CAAAJ63265-APEA 108.23 CAD
CAAAJ63265-AP CAAAJ63265-K2
Bisacrylamide 2% in aqueous solution
Bisacrylamide

Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis. Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels.


Liquid

Formula: (H₂C=CHCONH)₂CH₂
Storage Temperature: Ambient
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR