Indomethacin 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J63255-03
J63255-06
J63255-14
CAAAJ63255-03EA
39.03
CAD
CAAAJ63255-03
CAAAJ63255-06
CAAAJ63255-14
Indomethacin 99+%
Indomethacin
Non-selective cyclooxygenase inhibitor.
Powder
Formula:
C₁₉H₁₆ClNO₄ Melting Pt: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00057095 CAS Number: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: "6.1",III Merck Index: 14,4968 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...