Piroxicam

Supplier: Thermo Scientific Chemicals
Danger

J63239-03 J63239-06 J63239-09
CAAAJ63239-03EA 68.26 CAD
CAAAJ63239-03 CAAAJ63239-06 CAAAJ63239-09
Piroxicam
Piroxicam

A non-steroidal anti-inflammatory drug (NSAID) that is a highly selective inhibitor of cyclooxygenase-1 (COX-1)


Powder


Applications: A non-steroidal anti-inflammatory drug (NSAID) that is a highly selective inhibitor of cyclooxygenase-1 (COX-1)

Formula: C₁₅H₁₃N₃O₄S
Storage Temperature: Ambient
MDL Number: MFCD00057317
CAS Number: 36322-90-4
EINECS: 252-974-3
UN: 2811
ADR: "6.1",III
Merck Index: 14,7506

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR