Trioxsalen 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5',8-Trimethylpsoralen, Trisoralen, TMP, 2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one

J63226-03 J63226-06
CAAAJ63226-03EA 155.79 CAD
CAAAJ63226-03 CAAAJ63226-06
Trioxsalen 99%
Trioxsalen

Photochemical crosslinker of DNA. Useful for treatment of psoriasis


Powder


Applications: Photochemical crosslinker of DNA. Useful for treatment of psoriasis

Formula: C₁₄H₁₂O₃
Melting Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00005010
CAS Number: 3902-71-4
EINECS: 223-459-0
UN: 1759
ADR: 8,III
Merck Index: 14,9735

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR